Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRASP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271856.1 | NP_001258785.1 | 5 | 444 | ||
NM_181711.3 | NP_859062.1 | 6 | 678 | |||
XM_005268691.3 | XP_005268748.1 | 6 | 1846 | |||
XM_011537996.1 | XP_011536298.1 | 5 | 1624 | |||
GenBank mRNA | BC035500.2 | - | 5 | 415 | 4422992 | |
BC052967.1 | - | 5 | 362 | |||
BC053666.1 | - | 6 | 659 | 6151168 | ||
HY035281.1 | - | 5 | 378 |