Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 12, 13 | 1481 | ||
NM_001204505.2 | NP_001191434.1 | 9, 10 | 1102 | |||
NM_001204506.2 | NP_001191435.1 | 9, 10 | 1102 | |||
NM_001297429.1 | NP_001284358.1 | 7, 8 | 869 | |||
NM_152910.5 | NP_690874.2 | 11, 12 | 1444 | |||
NM_178009.4 | NP_821077.1 | 11, 12 | 1444 | |||
NR_123714.1 | 9, 10 | 1102 | ||||
NR_123715.1 | 11, 12 | 1715 | ||||
GenBank mRNA | AB078967.1 | - | 11, 12 | 1399 | ||
AB078968.1 | - | 11, 12 | 1399 | |||
AK122909.1 | - | 9, 10 | 1068 | |||
AK289546.1 | - | 12, 13 | 1483 | |||
AK302631.1 | - | 7, 8 | 829 | |||
AK302727.1 | - | 9, 10 | 1102 | |||
BC043292.2 | - | 9, 10 | 1059 | 5297895 |