Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204504.2 | NP_001191433.1 | 20 | 2438 | ||
NM_001204505.2 | NP_001191434.1 | 17 | 2059 | |||
NM_001204506.2 | NP_001191435.1 | 17 | 2059 | |||
NM_001297429.1 | NP_001284358.1 | 15 | 1826 | |||
NM_152910.5 | NP_690874.2 | 19 | 2401 | |||
NM_178009.4 | NP_821077.1 | 19 | 2401 | |||
NR_123714.1 | 17 | 2059 | ||||
NR_123715.1 | 20 | 2867 | ||||
GenBank mRNA | AB078967.1 | - | 19 | 2356 | ||
AB078968.1 | - | 19 | 2356 | |||
AK098302.1 | - | 4 | 420 | |||
AK122909.1 | - | 17 | 2024 | |||
AK289546.1 | - | 20 | 2440 | |||
AK302631.1 | - | 15 | 1786 | |||
AL833883.1 | - | 5 | 489 | |||
BC043292.2 | - | 17 | 2016 | 5297895 |