Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114309.1 | NP_001107781.1 | 3 | 578 | ||
NM_004433.4 | NP_004424.3 | 3 | 464 | |||
XM_005244942.2 | XP_005244999.1 | 3 | 507 | |||
GenBank mRNA | AF016295.1 | - | 3 | 425 | ||
AF017307.1 | - | 3 | 513 | |||
AF517841.1 | - | 3 | 446 | |||
AK225446.1 | - | 3 | 401 | |||
AK309265.1 | - | 2 | 272 | |||
AK310814.1 | - | 3 | 456 | |||
AK312273.1 | - | 3 | 440 | |||
BC003569.1 | - | 3 | 432 | 3537846 | ||
BX537368.1 | - | 2 | 3501 | |||
CR457106.1 | - | 2 | 311 | |||
CV570991.1 | - | 3 | 464 | |||
DA591406.1 | - | 3 | 554 | |||
U66894.1 | - | 3 | 406 | |||
U73843.1 | - | 3 | 430 | |||
U73844.1 | - | 3 | 430 | |||
U97156.1 | - | 3 | 435 |