Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114309.1 | NP_001107781.1 | 6 | 916 | ||
NM_004433.4 | NP_004424.3 | 6 | 802 | |||
XM_005244942.2 | XP_005244999.1 | 6 | 941 | |||
GenBank mRNA | AF016295.1 | - | 6 | 763 | ||
AF017307.1 | - | 6 | 851 | |||
AF517841.1 | - | 6 | 784 | |||
AK225446.1 | - | 6 | 739 | |||
AK309265.1 | - | 3 | 978 | |||
AK310814.1 | - | 6 | 794 | |||
AK312273.1 | - | 6 | 778 | |||
BC003569.1 | - | 6 | 770 | 3537846 | ||
BX537368.1 | - | 5 | 3839 | |||
CR457106.1 | - | 5 | 649 | |||
U66894.1 | - | 6 | 744 | |||
U73843.1 | - | 6 | 768 | |||
U73844.1 | - | 7 | 699 | |||
U97156.1 | - | 6 | 773 |