Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM229B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182526.2 | NP_872332.1 | 3 | 737 | ||
XM_005267373.2 | XP_005267430.1 | 3 | 743 | |||
XM_005267374.3 | XP_005267431.1 | 4 | 610 | |||
XM_005267375.2 | XP_005267432.1 | 3 | 802 | |||
XM_006720062.2 | XP_006720125.1 | 3 | 622 | |||
XM_017021051.1 | XP_016876540.1 | 4 | 894 | |||
XM_017021052.1 | XP_016876541.1 | 3 | 965 | |||
XM_017021053.1 | XP_016876542.1 | 3 | 869 | |||
XM_017021054.1 | XP_016876543.1 | 3 | 1107 | |||
XM_017021055.1 | XP_016876544.1 | 3 | 1029 | |||
GenBank mRNA | AK090706.1 | - | 3 | 559 | ||
BC104940.1 | - | 2 | 473 | 8143943 | ||
BC104942.1 | - | 2 | 473 | 8143945 |