Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM229B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182526.2 | NP_872332.1 | 3 | 1355 | ||
XM_005267373.2 | XP_005267430.1 | 3 | 1361 | |||
XM_005267374.3 | XP_005267431.1 | 4 | 1228 | |||
XM_005267375.2 | XP_005267432.1 | 3 | 1420 | |||
XM_006720062.2 | XP_006720125.1 | 3 | 1240 | |||
XM_017021051.1 | XP_016876540.1 | 4 | 1512 | |||
XM_017021052.1 | XP_016876541.1 | 3 | 1583 | |||
XM_017021053.1 | XP_016876542.1 | 3 | 1487 | |||
XM_017021054.1 | XP_016876543.1 | 3 | 1725 | |||
XM_017021055.1 | XP_016876544.1 | 3 | 1647 |