Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MDGA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113498.2 | NP_001106970.3 | 9 | 2108 | ||
NM_182830.4 | NP_878250.2 | 9 | 1814 | |||
XM_011536519.2 | XP_011534821.1 | 9 | 1963 | |||
XM_011536525.2 | XP_011534827.1 | 6 | 1245 | |||
XM_017021060.1 | XP_016876549.1 | 5 | 1074 | |||
GenBank mRNA | AK309211.1 | - | 9 | 2111 | ||
AY328482.1 | - | 3 | 741 | |||
AY358125.1 | - | 8 | 1688 | |||
AY369208.1 | - | 9 | 1904 | |||
BC142947.1 | - | 9 | 1831 |