Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOP9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286367.1 | NP_001273296.1 | 2 | 399 | ||
NM_174913.2 | NP_777573.1 | 2 | 399 | |||
XM_005267385.1 | XP_005267442.1 | 2 | 384 | |||
XM_011536526.1 | XP_011534828.1 | 2 | 176 | |||
XM_011536527.2 | XP_011534829.1 | 2 | 384 | |||
GenBank mRNA | AB095941.1 | - | 2 | 391 | ||
BX248018.1 | - | 2 | 361 | |||
HY061279.1 | - | 2 | 401 |