Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRED1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152594.2 | NP_689807.1 | 7 | 1511 | ||
XM_005254202.3 | XP_005254259.1 | 8 | 2311 | |||
XM_011521289.2 | XP_011519591.1 | 7 | 1208 | |||
GenBank mRNA | AK091222.1 | - | 8 | 1281 | ||
AK095219.1 | - | 1 | 105 | |||
AY299089.1 | - | 7 | 1511 | |||
BC137480.1 | - | 7 | 2036 | 9021480 | ||
BC137481.1 | - | 7 | 2036 | 9021481 |