Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FSIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324338.1 | NP_001311267.1 | 11 | 1463 | ||
NM_152597.4 | NP_689810.3 | 11 | 1560 | |||
XM_011521305.2 | XP_011519607.1 | 11 | 1436 | |||
XM_011521306.1 | XP_011519608.1 | 11 | 1849 | |||
XM_011521311.2 | XP_011519613.1 | 7 | 2120 | |||
XM_017021972.1 | XP_016877461.1 | 7 | 913 | |||
GenBank mRNA | AK093308.1 | - | 11 | 1542 | ||
AY260140.1 | - | 10 | 1357 | |||
BC045191.2 | - | 12 | 1552 | 5297527 |