Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITPRIPL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034841.3 | NP_001030013.1 | 1 | 841 | ||
NR_028028.1 | 2 | 325 | ||||
GenBank mRNA | AK096327.1 | - | 2 | 325 | ||
AK097274.1 | - | 1 | 287 | |||
BC101749.1 | - | 1 | 390 | 8069255 | ||
BC101751.1 | - | 1 | 390 | 8069257 | ||
BX335323.2 | - | 1 | 790 | |||
BX338638.2 | - | 1 | 783 | |||
BX382445.2 | - | 1 | 819 |