Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318469.1 | NP_001305398.1 | 4 | 623 | ||
NM_001318471.1 | NP_001305400.1 | 5 | 679 | |||
NM_001318472.1 | NP_001305401.1 | 3 | 546 | |||
NM_001318473.1 | NP_001305402.1 | 3 | 496 | |||
NM_001318474.1 | NP_001305403.1 | 3 | 477 | |||
NM_001318475.1 | NP_001305404.1 | 4 | 635 | |||
NM_001318476.1 | NP_001305405.1 | 4 | 621 | |||
NM_153688.3 | NP_710155.2 | 4 | 604 | |||
XM_011522921.2 | XP_011521223.1 | 6 | 664 | |||
XM_017022983.1 | XP_016878472.1 | 4 | 791 | |||
XM_017022984.1 | XP_016878473.1 | 5 | 918 | |||
XM_017022985.1 | XP_016878474.1 | 5 | 923 | |||
XM_017022986.1 | XP_016878475.1 | 6 | 1011 | |||
XM_017022987.1 | XP_016878476.1 | 4 | 635 | |||
GenBank mRNA | AK094761.1 | - | 4 | 541 | ||
AK291372.1 | - | 3 | 424 | |||
AK296560.1 | - | 4 | 553 | |||
AK308134.1 | - | 3 | 453 | |||
AL038510.1 | - | 4 | 522 | |||
AL538214.3 | - | 4 | 491 | |||
BC064604.1 | - | 4 | 482 | |||
BC113863.1 | - | 2 | 295 | |||
BC114471.1 | - | 2 | 295 | |||
BC127184.1 | - | 2 | 295 | |||
BX647144.1 | - | 2 | 259 | |||
DA781502.1 | - | 3 | 400 | |||
DC371394.1 | - | 3 | 424 |