Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318469.1 | NP_001305398.1 | 4 | 1364 | ||
NM_001318471.1 | NP_001305400.1 | 5 | 1420 | |||
NM_001318472.1 | NP_001305401.1 | 3 | 1287 | |||
NM_001318473.1 | NP_001305402.1 | 3 | 1237 | |||
NM_001318474.1 | NP_001305403.1 | 3 | 1218 | |||
NM_001318475.1 | NP_001305404.1 | 4 | 1376 | |||
NM_001318476.1 | NP_001305405.1 | 4 | 1362 | |||
NM_153688.3 | NP_710155.2 | 4 | 1345 | |||
XM_011522921.2 | XP_011521223.1 | 6 | 1405 | |||
XM_017022983.1 | XP_016878472.1 | 4 | 1532 | |||
XM_017022984.1 | XP_016878473.1 | 5 | 1659 | |||
XM_017022985.1 | XP_016878474.1 | 5 | 1664 | |||
XM_017022986.1 | XP_016878475.1 | 6 | 1752 | |||
XM_017022987.1 | XP_016878476.1 | 4 | 1376 | |||
GenBank mRNA | AK091562.1 | - | 1 | 572 | ||
AK094761.1 | - | 4 | 1282 | |||
AK291372.1 | - | 3 | 1165 | |||
AK296560.1 | - | 4 | 1294 | |||
AK308134.1 | - | 3 | 1193 | |||
BC033774.1 | - | 1 | 708 | |||
BC064604.1 | - | 4 | 1223 | |||
BC113863.1 | - | 2 | 1036 | |||
BC114471.1 | - | 2 | 1036 | |||
BC127184.1 | - | 2 | 1036 | |||
BX647144.1 | - | 2 | 1000 |