Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHBDL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138328.2 | NP_612201.1 | 2, 3 | 142 | ||
XM_017024272.1 | XP_016879761.1 | 2, 3 | 147 | |||
GenBank mRNA | AJ313480.1 | - | 2, 3 | 142 | ||
BC100975.1 | - | 2, 3 | 141 | 40005244 | ||
BC100976.1 | - | 2, 3 | 142 |