Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF433 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080411.2 | NP_001073880.1 | 4 | 827 | ||
NM_001308346.1 | NP_001295275.1 | 5 | 886 | |||
NM_001308348.1 | NP_001295277.1 | 4 | 818 | |||
NM_001308351.1 | NP_001295280.1 | 6 | 924 | |||
NM_001308355.1 | NP_001295284.1 | 5 | 877 | |||
NM_001308357.1 | NP_001295286.1 | 5 | 865 | |||
XM_006722661.3 | XP_006722724.1 | 4 | 726 | |||
XM_011527725.2 | XP_011526027.1 | 4 | 788 | |||
XM_011527728.2 | XP_011526030.1 | 6 | 814 | |||
XM_011527729.2 | XP_011526031.1 | 5 | 755 | |||
XM_017026413.1 | XP_016881902.1 | 3 | 1358 | |||
GenBank mRNA | AK098300.1 | - | 4 | 796 | ||
BC047412.1 | - | 5 | 800 | 5295132 |