Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 16, 17 | 1870 | ||
NM_004434.2 | NP_004425.2 | 15, 16 | 1813 | |||
XM_005267397.1 | XP_005267454.1 | 16, 17 | 1940 | |||
XM_005267398.3 | XP_005267455.1 | 16, 17 | 1873 | |||
XM_005267399.4 | XP_005267456.2 | 16, 17 | 1912 | |||
XM_005267400.4 | XP_005267457.1 | 15, 16 | 1839 | |||
XM_011536540.2 | XP_011534842.1 | 15, 16 | 1886 | |||
XM_011536541.1 | XP_011534843.1 | 16, 17 | 1953 | |||
GenBank mRNA | AK126980.1 | - | 17, 18 | 1826 | ||
AK299812.1 | - | 16, 17 | 2149 | |||
BC032541.1 | - | 15, 16 | 1789 | 5533599 | ||
BC033043.1 | - | 15, 16 | 1813 | 4821202 | ||
BX442035.2 | - | 3, 4 | 369 | |||
U97018.1 | - | 15, 16 | 1812 |