Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152604.1 | NP_689817.1 | 5 | 1063 | ||
XM_005258585.2 | XP_005258642.1 | 6 | 1321 | |||
XM_005258587.2 | XP_005258644.1 | 5 | 1198 | |||
XM_005258588.2 | XP_005258645.1 | 6 | 960 | |||
XM_011526586.2 | XP_011524888.1 | 7 | 2066 | |||
XM_011526587.2 | XP_011524889.1 | 6 | 2033 | |||
XM_011526588.2 | XP_011524890.1 | 5 | 2215 | |||
XM_011526589.2 | XP_011524891.1 | 5 | 896 | |||
XM_011526590.2 | XP_011524892.1 | 4 | 861 | |||
XM_017026422.1 | XP_016881911.1 | 7 | 1472 | |||
XM_017026423.1 | XP_016881912.1 | 7 | 2066 | |||
XM_017026424.1 | XP_016881913.1 | 5 | 1112 | |||
GenBank mRNA | AK091760.1 | - | 5 | 1624 | ||
AK093182.1 | - | 5 | 1063 | |||
AK122892.1 | - | 8 | 1389 | |||
AY260141.1 | - | 4 | 858 | |||
AY438646.1 | - | 5 | 1063 |