Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF780B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005851.2 | NP_001005851.1 | 4 | 1611 | ||
XM_005258590.4 | XP_005258647.1 | 6 | 1843 | |||
XM_005258591.4 | XP_005258648.1 | 5 | 4432 | |||
XM_005258592.3 | XP_005258649.1 | 6 | 1751 | |||
XM_005258593.2 | XP_005258650.1 | 6 | 1827 | |||
XM_005258595.3 | XP_005258652.1 | 5 | 1677 | |||
XM_006723072.3 | XP_006723135.1 | 6 | 4593 | |||
XM_011526593.2 | XP_011524895.1 | 5 | 1623 | |||
XM_017026425.1 | XP_016881914.1 | 6 | 4590 | |||
XM_017026426.1 | XP_016881915.1 | 6 | 1695 | |||
XM_017026427.1 | XP_016881916.1 | 5 | 1674 | |||
GenBank mRNA | AK127063.1 | - | 6 | 1804 | ||
BC136960.1 | - | 4 | 1591 | 9020952 |