Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 20 | 2168 | ||
NM_004434.2 | NP_004425.2 | 19 | 2111 | |||
XM_005267397.1 | XP_005267454.1 | 20 | 2238 | |||
XM_005267398.3 | XP_005267455.1 | 20 | 2171 | |||
XM_005267399.4 | XP_005267456.2 | 20 | 2210 | |||
XM_005267400.4 | XP_005267457.1 | 19 | 2137 | |||
XM_011536540.2 | XP_011534842.1 | 19 | 2184 | |||
XM_011536541.1 | XP_011534843.1 | 20 | 2251 | |||
XM_011536542.2 | XP_011534844.1 | 19 | 2078 | |||
XM_017021074.1 | XP_016876563.1 | 18 | 2049 | |||
GenBank mRNA | AK126980.1 | - | 21 | 2124 | ||
AK299812.1 | - | 20 | 2447 | |||
BC032541.1 | - | 19 | 2087 | 5533599 | ||
BC033043.1 | - | 19 | 2111 | 4821202 | ||
BX442035.2 | - | 7 | 667 | |||
U97018.1 | - | 19 | 2110 |