Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 14 | 1604 | ||
NM_004434.2 | NP_004425.2 | 13 | 1547 | |||
XM_005267397.1 | XP_005267454.1 | 14 | 1674 | |||
XM_005267398.3 | XP_005267455.1 | 14 | 1607 | |||
XM_005267399.4 | XP_005267456.2 | 14 | 1646 | |||
XM_005267400.4 | XP_005267457.1 | 13 | 1573 | |||
XM_011536540.2 | XP_011534842.1 | 13 | 1620 | |||
XM_011536541.1 | XP_011534843.1 | 14 | 1687 | |||
XM_011536542.2 | XP_011534844.1 | 14 | 1646 | |||
XM_017021074.1 | XP_016876563.1 | 13 | 1617 | |||
GenBank mRNA | AK023861.1 | - | 4 | 1236 | ||
AK126980.1 | - | 15 | 1560 | |||
AK299812.1 | - | 14 | 1883 | |||
BC032541.1 | - | 13 | 1523 | 5533599 | ||
BC033043.1 | - | 13 | 1547 | 4821202 | ||
BX247979.1 | - | 14 | 1616 | |||
BX442035.2 | - | 1 | 103 | |||
U97018.1 | - | 13 | 1546 |