Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256404.1 | NP_001243333.1 | 8 | 2015 | ||
NM_198459.3 | NP_940861.3 | 5 | 1475 | |||
GenBank mRNA | AK308302.1 | - | 6 | 1666 | ||
BX647950.1 | - | 5 | 1476 | |||
BX649075.1 | - | 5 | 601 | |||
CR749576.1 | - | 8 | 1939 |