Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_170743.3 | NP_734464.1 | 5 | 640 | ||
NM_173064.2 | NP_775087.1 | 5 | 640 | |||
NM_173065.2 | NP_775088.1 | 5 | 640 | |||
XM_006710394.3 | XP_006710457.2 | 4 | 704 | |||
XM_017000479.1 | XP_016855968.1 | 4 | 669 | |||
GenBank mRNA | AF439325.1 | - | 5 | 613 | ||
AJ534330.1 | - | 5 | 626 | |||
AJ534331.1 | - | 5 | 626 | |||
AK160364.1 | - | 5 | 616 | |||
AY129151.1 | - | 5 | 613 | |||
AY129152.1 | - | 5 | 613 | |||
BC101407.1 | - | 4 | 502 | |||
BC101408.1 | - | 4 | 502 | |||
BC101975.1 | - | 5 | 626 | |||
BC139722.1 | - | 5 | 626 | |||
BC140872.1 | - | 5 | 689 | 9021740 | ||
JN591376.1 | - | 5 | 613 |