Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MARK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039469.2 | NP_001034558.2 | 13 | 1865 | ||
NM_001163296.1 | NP_001156768.1 | 13 | 1865 | |||
NM_001163297.1 | NP_001156769.1 | 13 | 1862 | |||
NM_004954.4 | NP_004945.4 | 13 | 1862 | |||
NM_017490.3 | NP_059672.2 | 13 | 1679 | |||
XM_011544789.1 | XP_011543091.1 | 14 | 1756 | |||
XM_011544790.1 | XP_011543092.1 | 14 | 1756 | |||
XM_011544791.1 | XP_011543093.1 | 14 | 1753 | |||
XM_011544792.1 | XP_011543094.1 | 14 | 1756 | |||
XM_011544793.1 | XP_011543095.1 | 14 | 1756 | |||
XM_011544794.1 | XP_011543096.1 | 14 | 1753 | |||
XM_011544795.1 | XP_011543097.1 | 14 | 1756 | |||
XM_011544796.1 | XP_011543098.1 | 14 | 1753 | |||
XM_011544797.1 | XP_011543099.1 | 14 | 1756 | |||
XM_011544798.1 | XP_011543100.1 | 14 | 1753 | |||
XM_017017310.1 | XP_016872799.1 | 13 | 1835 | |||
XM_017017311.1 | XP_016872800.1 | 14 | 1753 | |||
GenBank mRNA | AB188493.1 | - | 13 | 1869 | ||
AB271694.1 | - | 12 | 1184 | |||
AF387638.1 | - | 12 | 1202 | |||
AK225766.1 | - | 13 | 1697 | |||
AK290339.1 | - | 13 | 1647 | |||
BC008771.2 | - | 13 | 1257 | |||
BC084540.1 | - | 13 | 1321 | 4591688 | ||
BT007342.1 | - | 12 | 1187 | |||
X97630.1 | - | 13 | 1591 |