Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105669.2 | NP_001099139.2 | 10 | 1675 | ||
XM_011509253.2 | XP_011507555.1 | 9 | 1057 | |||
XM_011509254.2 | XP_011507556.1 | 9 | 1054 | |||
XM_011509255.2 | XP_011507557.1 | 8 | 943 | |||
XM_017000497.1 | XP_016855986.1 | 7 | 679 |