Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBL4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_203412.1 | NP_981957.1 | 1 | 680 | ||
GenBank mRNA | AK098556.1 | - | 1 | 681 | ||
BC058929.1 | - | 1 | 652 | 5741847 |