Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152611.4 | NP_689824.2 | 5 | 1662 | ||
XM_011529183.2 | XP_011527485.1 | 2 | 642 | |||
GenBank mRNA | AK290123.1 | - | 5 | 1575 | ||
BC019612.1 | - | 1 | 474 | |||
BC027720.2 | - | 5 | 1612 | 4588586 |