Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001423.2 | NP_001414.1 | 2, 3 | 334 | ||
GenBank mRNA | AK299388.1 | - | 2, 3 | 216 | ||
AK312246.1 | - | 2, 3 | 208 | |||
BC047300.1 | - | 2, 3 | 259 | 4444217 | ||
DC372133.1 | - | 2, 3 | 468 | |||
U43916.1 | - | 1, 2 | 87 | |||
U77085.1 | - | 2, 3 | 303 | |||
Y07909.1 | - | 2, 3 | 305 | |||
Z50751.1 | - | 2, 3 | 209 |