Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APOBEC3H | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001166002.2 | NP_001159474.2 | 3 | 358 | ||
NM_001166003.2 | NP_001159475.2 | 3 | 358 | |||
NM_001166004.2 | NP_001159476.2 | 3 | 358 | |||
NM_181773.4 | NP_861438.3 | 3 | 358 | |||
XM_011529990.2 | XP_011528292.1 | 3 | 378 | |||
XM_011529991.2 | XP_011528293.1 | 3 | 378 | |||
XM_011529992.2 | XP_011528294.1 | 3 | 377 | |||
GenBank mRNA | BC069023.1 | - | 3 | 343 | 5441646 | |
BQ052182.1 | - | 3 | 297 | |||
BQ054330.1 | - | 3 | 304 | |||
BU585251.1 | - | 3 | 359 | |||
CR456481.3 | - | 3 | 271 | |||
FJ376611.1 | - | 2 | 231 | |||
FJ376612.1 | - | 2 | 231 | |||
FJ376613.1 | - | 2 | 231 | |||
FJ376614.1 | - | 2 | 231 | |||
FJ376615.1 | - | 2 | 231 | |||
FJ376616.1 | - | 2 | 231 | |||
FJ376617.1 | - | 2 | 231 | |||
KC329529.1 | - | 2 | 231 |