Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145168.1 | NP_001138640.1 | 12, 13 | 3077 | ||
NM_001145169.1 | NP_001138641.1 | 11, 12 | 2923 | |||
NM_001321971.1 | NP_001308900.1 | 11, 12 | 3456 | |||
NM_001321975.1 | NP_001308904.1 | 12, 13 | 2997 | |||
NM_153835.3 | NP_722577.2 | 9, 10 | 3063 | |||
NR_135916.1 | 9, 10 | 3063 | ||||
XM_011532621.2 | XP_011530923.1 | 11, 12 | 3408 | |||
XM_011532622.2 | XP_011530924.1 | 11, 12 | 3360 | |||
XM_011532623.2 | XP_011530925.1 | 11, 12 | 3352 | |||
XM_017003505.1 | XP_016858994.1 | 11, 12 | 3394 | |||
XM_017003506.1 | XP_016858995.1 | 11, 12 | 3650 | |||
GenBank mRNA | AK122995.1 | - | 6, 7 | 2568 | ||
AK293887.1 | - | 11, 12 | 2923 | |||
AY140955.1 | - | 12, 13 | 3077 | |||
AY358172.1 | - | 9, 10 | 3063 | |||
AY358214.1 | - | 3, 4 | 1534 |