Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145168.1 | NP_001138640.1 | 11 | 1943 | ||
NM_001145169.1 | NP_001138641.1 | 10 | 1789 | |||
NM_001321971.1 | NP_001308900.1 | 10 | 2322 | |||
NM_001321975.1 | NP_001308904.1 | 11 | 1863 | |||
NM_153835.3 | NP_722577.2 | 8 | 1929 | |||
NR_135916.1 | 8 | 1929 | ||||
XM_011532621.2 | XP_011530923.1 | 10 | 2274 | |||
XM_011532622.2 | XP_011530924.1 | 10 | 2226 | |||
XM_011532623.2 | XP_011530925.1 | 10 | 2218 | |||
XM_017003505.1 | XP_016858994.1 | 10 | 2260 | |||
XM_017003506.1 | XP_016858995.1 | 10 | 2516 | |||
GenBank mRNA | AK122995.1 | - | 5 | 1434 | ||
AK293887.1 | - | 10 | 1789 | |||
AY140955.1 | - | 11 | 1943 | |||
AY358172.1 | - | 8 | 1929 |