Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM179A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199280.2 | NP_954974.2 | 10, 11 | 1712 | ||
XM_011532628.2 | XP_011530930.1 | 10, 11 | 1712 | |||
XM_017003507.1 | XP_016858996.1 | 10, 11 | 1761 | |||
XM_017003508.1 | XP_016858997.1 | 7, 8 | 1394 | |||
XM_017003509.1 | XP_016858998.1 | 5, 6 | 685 | |||
XM_017003510.1 | XP_016858999.1 | 3, 4 | 450 | |||
XM_017003511.1 | XP_016859000.1 | 4, 5 | 483 | |||
XM_017003512.1 | XP_016859001.1 | 10, 11 | 1712 |