Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM171B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177454.3 | NP_803237.3 | 8 | 2420 | ||
GenBank mRNA | AB075826.1 | - | 7 | 1869 | ||
AK091423.1 | - | 9 | 2325 | |||
AK094578.1 | - | 1 | 828 | |||
AK095604.1 | - | 5 | 1702 | |||
AK289737.1 | - | 8 | 2423 | |||
AL834264.1 | - | 1 | 822 | |||
BC060872.1 | - | 8 | 2426 | 30347998 |