Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PARP15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113523.2 | NP_001106995.1 | 11 | 1787 | ||
NM_001308320.1 | NP_001295249.1 | 8 | 1202 | |||
NM_001308321.1 | NP_001295250.1 | 6 | 907 | |||
NM_152615.2 | NP_689828.1 | 7 | 1114 | |||
XM_005247159.3 | XP_005247216.1 | 10 | 3252 | |||
XM_005247160.3 | XP_005247217.1 | 10 | 3231 | |||
XM_011512475.2 | XP_011510777.1 | 10 | 3343 | |||
XM_011512476.2 | XP_011510778.1 | 10 | 3312 | |||
XM_011512477.2 | XP_011510779.1 | 9 | 3081 | |||
XM_011512478.2 | XP_011510780.1 | 11 | 1570 | |||
XM_011512479.2 | XP_011510781.1 | 9 | 1045 | |||
XM_017005791.1 | XP_016861280.1 | 9 | 3024 | |||
XM_017005792.1 | XP_016861281.1 | 8 | 2212 | |||
GenBank mRNA | AK097515.1 | - | 13 | 1732 | ||
AK097916.1 | - | 7 | 1051 | |||
BC101701.1 | - | 7 | 1114 | 8069207 | ||
BC101703.1 | - | 7 | 1114 | 8069209 | ||
BC143573.1 | - | 6 | 907 | 9052084 | ||
BX647656.1 | - | 8 | 1178 | |||
DQ063586.1 | - | 11 | 1706 |