Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207335.2 | NP_997218.2 | 5 | 1712 | ||
XM_005247167.1 | XP_005247224.1 | 6 | 2176 | |||
XM_005247168.4 | XP_005247225.1 | 6 | 2254 | |||
XM_011512490.2 | XP_011510792.1 | 6 | 2176 | |||
GenBank mRNA | AK128176.1 | - | 5 | 574 | ||
AK307108.1 | - | 3 | 329 | |||
BC041365.1 | - | 2 | 71 | |||
BX537865.1 | - | 5 | 751 |