Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KBTBD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207335.2 | NP_997218.2 | 3 | 1471 | ||
XM_005247167.1 | XP_005247224.1 | 4 | 1935 | |||
XM_005247168.4 | XP_005247225.1 | 4 | 2013 | |||
XM_011512490.2 | XP_011510792.1 | 4 | 1935 | |||
XM_011512491.2 | XP_011510793.1 | 4 | 1935 | |||
XM_011512492.2 | XP_011510794.1 | 4 | 1935 | |||
GenBank mRNA | AK128176.1 | - | 3 | 333 | ||
BX537865.1 | - | 3 | 510 |