Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317799.1 | NP_001304728.1 | 2 | 339 | ||
NM_145207.2 | NP_660208.2 | 2 | 342 | |||
XM_005262783.4 | XP_005262840.1 | 2 | 339 | |||
XM_011531678.2 | XP_011529980.1 | 2 | 339 | |||
XM_011531679.2 | XP_011529981.1 | 2 | 297 | |||
XM_017007825.1 | XP_016863314.1 | 2 | 338 | |||
XM_017007826.1 | XP_016863315.1 | 2 | 338 | |||
XM_017007827.1 | XP_016863316.1 | 2 | 338 | |||
XM_017007828.1 | XP_016863317.1 | 2 | 304 | |||
XM_017007830.1 | XP_016863319.1 | 2 | 338 | |||
GenBank mRNA | AF361489.1 | - | 2 | 338 | ||
AF479656.1 | - | 2 | 312 | |||
AK091384.1 | - | 2 | 297 | |||
AL832175.1 | - | 2 | 335 | |||
BC048217.1 | - | 2 | 282 | 6052305 | ||
DA619323.1 | - | 2 | 340 |