Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004097.2 | NP_004088.2 | 3 | 1108 | ||
XM_011532697.2 | XP_011530999.1 | 3 | 6128 | |||
GenBank mRNA | AK092368.1 | - | 1 | 640 | ||
BC037242.1 | - | 3 | 975 | |||
BC045762.1 | - | 3 | 1108 | 5260039 | ||
X68879.1 | - | 3 | 319 |