Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SGMS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136257.1 | NP_001129729.1 | 2 | 451 | ||
NM_001136258.1 | NP_001129730.1 | 3 | 643 | |||
NM_152621.5 | NP_689834.1 | 2 | 727 | |||
XM_011531698.2 | XP_011530000.1 | 3 | 781 | |||
XM_011531699.2 | XP_011530001.1 | 4 | 655 | |||
XM_011531700.2 | XP_011530002.1 | 3 | 552 | |||
XM_011531701.2 | XP_011530003.1 | 4 | 1666 | |||
XM_011531702.2 | XP_011530004.1 | 3 | 1594 | |||
XM_017007839.1 | XP_016863328.1 | 3 | 785 | |||
XM_017007840.1 | XP_016863329.1 | 4 | 907 | |||
XM_017007841.1 | XP_016863330.1 | 3 | 781 | |||
GenBank mRNA | AF452717.1 | - | 3 | 546 | ||
AK290344.1 | - | 3 | 645 | |||
AK314049.1 | - | 2 | 442 | |||
BC028705.1 | - | 2 | 452 | 4823252 | ||
BC041369.2 | - | 2 | 720 | 5273650 | ||
BX649142.1 | - | 1 | 108 |