Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM151B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_205548.2 | NP_991111.2 | 4 | 537 | ||
XM_006714564.3 | XP_006714627.2 | 4 | 455 | |||
XM_011543234.2 | XP_011541536.1 | 4 | 916 | |||
XM_011543235.2 | XP_011541537.1 | 3 | 291 | |||
XM_011543236.2 | XP_011541538.1 | 4 | 667 | |||
XM_011543237.2 | XP_011541539.1 | 3 | 838 | |||
XM_017009166.1 | XP_016864655.1 | 4 | 854 | |||
XM_017009167.1 | XP_016864656.1 | 4 | 864 | |||
XM_017009168.1 | XP_016864657.1 | 5 | 583 | |||
XM_017009169.1 | XP_016864658.1 | 3 | 363 | |||
XM_017009170.1 | XP_016864659.1 | 3 | 788 | |||
GenBank mRNA | AY358256.1 | - | 4 | 537 | ||
BC131563.1 | - | 4 | 469 | 40109365 | ||
BC142990.1 | - | 9 | 1538 |