Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF92 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287532.1 | NP_001274461.1 | 3 | 445 | ||
NM_001287533.1 | NP_001274462.1 | 5 | 663 | |||
NM_001287534.1 | NP_001274463.1 | 2 | 318 | |||
NM_007139.3 | NP_009070.2 | 3 | 414 | |||
NM_152626.3 | NP_689839.1 | 4 | 541 | |||
GenBank mRNA | AK091618.1 | - | 4 | 459 | ||
BC036439.1 | - | 3 | 337 | 4817733 | ||
BC040594.1 | - | 4 | 483 | 5269950 | ||
BX647875.1 | - | 2 | 263 | |||
DA397874.1 | - | 3 | 301 | |||
DA637374.1 | - | 2 | 278 | |||
DA939471.1 | - | 3 | 387 | |||
EU668348.1 | - | 4 | 485 | |||
M61872.1 | - | 3 | 339 |