Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BMPER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133468.4 | NP_597725.1 | 14 | 1882 | ||
XM_005249633.2 | XP_005249690.1 | 12 | 1292 | |||
XM_017011800.1 | XP_016867289.1 | 12 | 1442 | |||
XM_017011801.1 | XP_016867290.1 | 11 | 1112 | |||
GenBank mRNA | AB075845.1 | - | 10 | 1149 | ||
AK289963.1 | - | 13 | 1622 | |||
AY324650.1 | - | 14 | 1881 | |||
BC060868.1 | - | 13 | 1559 | 30345290 |