Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176814.4 | NP_789784.2 | 3, 4 | 878 | ||
XM_005250181.3 | XP_005250238.1 | 3, 4 | 952 | |||
XM_006715867.3 | XP_006715930.1 | 3, 4 | 957 | |||
XM_006715868.3 | XP_006715931.1 | 3, 4 | 952 | |||
XM_011515859.2 | XP_011514161.1 | 3, 4 | 673 | |||
GenBank mRNA | AF218032.1 | - | 3, 4 | 848 | ||
AK297946.1 | - | 1, 2 | 66 | |||
AK309674.1 | - | 3, 4 | 872 | |||
BC047641.1 | - | 3, 4 | 897 | |||
BC110623.1 | - | 3, 4 | 423 | 40036036 | ||
BC110624.1 | - | 3, 4 | 418 | 40036038 | ||
DQ655936.2 | - | 3, 4 | 269 |