Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL22A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152888.2 | NP_690848.1 | 8 | 1734 | ||
XM_011516883.2 | XP_011515185.1 | 8 | 1812 | |||
XM_011516884.2 | XP_011515186.1 | 8 | 1812 | |||
XM_011516885.2 | XP_011515187.1 | 8 | 1812 | |||
XM_011516886.2 | XP_011515188.1 | 9 | 1724 | |||
XM_011516887.2 | XP_011515189.1 | 5 | 692 | |||
XM_011516888.2 | XP_011515190.1 | 8 | 1815 | |||
XM_017013150.1 | XP_016868639.1 | 9 | 1685 | |||
XM_017013151.1 | XP_016868640.1 | 8 | 1812 | |||
XM_017013152.1 | XP_016868641.1 | 4 | 682 | |||
GenBank mRNA | AF406780.1 | - | 8 | 1721 |