Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152628.3 | NP_689841.3 | 12, 13 | 1329 | ||
XM_005250815.3 | XP_005250872.1 | 9, 10 | 968 | |||
XM_011516899.1 | XP_011515201.1 | 11, 12 | 1306 | |||
XM_017013153.1 | XP_016868642.1 | 12, 13 | 1329 | |||
XM_017013157.1 | XP_016868646.1 | 11, 12 | 1215 | |||
XM_017013160.1 | XP_016868649.1 | 11, 12 | 1125 | |||
XM_017013161.1 | XP_016868650.1 | 9, 10 | 1080 | |||
XM_017013162.1 | XP_016868651.1 | 10, 11 | 1011 | |||
XM_017013163.1 | XP_016868652.1 | 9, 10 | 1409 | |||
GenBank mRNA | AK093906.1 | - | 12, 13 | 1329 | ||
BC031260.1 | - | 9, 10 | 920 | 5272635 | ||
CD106024.1 | - | 7, 8 | 740 |