Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM64 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008495.3 | NP_001008495.2 | 1, 2 | 788 | ||
NM_001146273.1 | NP_001139745.1 | 1, 2 | 788 | |||
GenBank mRNA | AK123958.1 | - | 1, 2 | 482 | ||
AK300583.1 | - | 1, 2 | 395 | |||
AL834364.1 | - | 1, 2 | 494 | |||
AY147881.1 | - | 1, 2 | 593 |