Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF596 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042415.2 | NP_001035880.1 | 6 | 697 | ||
NM_001042416.2 | NP_001035881.1 | 6 | 830 | |||
NM_001287254.1 | NP_001274183.1 | 6 | 688 | |||
NM_001287255.1 | NP_001274184.1 | 6 | 663 | |||
NM_001287256.1 | NP_001274185.1 | 6 | 659 | |||
NM_001287399.1 | NP_001274328.1 | 6 | 746 | |||
NM_173539.3 | NP_775810.2 | 6 | 748 | |||
XM_011534744.1 | XP_011533046.1 | 6 | 670 | |||
XM_017013166.1 | XP_016868655.1 | 5 | 2387 | |||
GenBank mRNA | AK313454.1 | - | 6 | 571 | ||
BC026190.1 | - | 6 | 571 | 4837032 | ||
BG771504.1 | - | 6 | 572 | |||
BM478564.1 | - | 6 | 559 |