Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM46D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170574.1 | NP_001164045.1 | 5 | 1336 | ||
NM_152630.4 | NP_689843.1 | 3 | 1260 | |||
GenBank mRNA | AK128147.1 | - | 5 | 1336 | ||
AK313875.1 | - | 3 | 1231 | |||
BC028710.2 | - | 3 | 1229 | 4827590 | ||
BC030784.2 | - | 3 | 1241 | 4826421 | ||
BC034979.2 | - | 3 | 1226 | 4824369 | ||
BX537938.1 | - | 3 | 1262 |