Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139058.2 | NP_620689.1 | 5 | 1963 | ||
GenBank mRNA | BQ269551.1 | - | 1 | 151 | ||
CA775911.1 | - | 1 | 142 |