Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OIT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152635.2 | NP_689848.1 | 2 | 598 | ||
NR_130125.1 | 2 | 598 | ||||
GenBank mRNA | AK096435.1 | - | 2 | 573 | ||
AY013707.2 | - | 2 | 429 | |||
AY358339.1 | - | 2 | 551 | |||
BC126440.1 | - | 2 | 570 | 8992156 |